Learn More
G-Biosciences™ Well-Coated™ Nickel Plate
SCI_ED
Catalog No. 501117990
Change view
Click to view available options
Color:
Clear
Catalog No. Color
50-111-7990 Clear
1 options
Catalog No. 50-111-7990 Supplier G-Biosciences™ Supplier No. 786749
Add to Cart
Add to Cart
96-Well Plates coated with nickel for binding 6X His Tagged Proteins.
Well-Coated™ Nickel plates are designed to specifically bind 6X histidine (polyhistidine) tagged proteins and peptides. The plates can be used for the isolation of polyhistidine-tagged proteins direct from bacterial lysates and then used for subsequent ELISA protocols.
  • Activated to 200 μL depth
  • Low non-specific binding
  • Ideal for proteins and peptides with polyhistidine (6X His) tag
  • Reduced non-specific binding as plates are pre-blocked with Superior™ Blocking Buffer
Specifications
Description Strip Plate
Color Clear
Quantity 5 x 8-well plates