Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-Biosciences™ Well-Coated™ Streptavidin Plate
SCI_ED

Catalog No. 501035877
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
50-103-5877 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-103-5877 Supplier G-Biosciences™ Supplier No. 786779
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

96-Well Plates coated with streptavidin for binding Biotinylated Molecules.

Well-Coated™ Streptavidin plates are designed to specifically bind biotinylated molecules, including biotin tagged antibodies. This is particular advantageous for antibodies known to denature upon direct binding to polystyrene plates.

  • High binding capacity for biotin
  • Low non-specific binding
  • Ideal for peptides, antibodies and small hydrophilic molecules
  • Reduced non-specific binding as plates are pre-blocked with Superior™ Blocking Buffer

Specifications

Content And Storage Upon arrival, store unopened at 4°C
Description Strip Plate
Color White
Quantity 5 x 8-well plates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.