Learn More
Chemglass Life Sciences Well Locator, Plastic, White, Reusable
SDP
Catalog No. 501532317
Encompass
Well Locator, Plastic, White, Reusable. Place this 96 well alpha-numeric printed locator under your 96 well plate for quick and simple well orientation. Ideal for ELISA or cell culture work. Reusable Template with Each Well Address Printed in Bold Letters. Fits Under Multi-well Plate. Makes Identifying Individual Wells Effortless. Stress Free Sample Retrieval. Best Used with Well Locator Stand (CLS-3478)
Catalog No. 50-153-2317 Supplier Chemglass Life Sciences Supplier No. CLS3477W01
Add to Cart
Add to Cart
Well Locator, Plastic, White, Reusable. Place this 96 well alpha-numeric printed locator under your 96 well plate for quick and simple well orientation. Ideal for ELISA or cell culture work. Reusable Template with Each Well Address Printed in Bold Letters. Fits Under Multi-well Plate. Makes Identifying Individual Wells Effortless. Stress Free Sample Retrieval. Best Used with Well Locator Stand (CLS-3478)