Learn More
Chemglass Life Sciences Well Locator Stand
SDP
Catalog No. 501532318
Encompass
Well Locator Stand. Works perfectly with printed well-orienters. Place your 96 well plate on this 30 angled platform for easier visibility. Convenient Stand to be Used with CLS-3477 Well Locators. Tilted at a 30 Angle for Easy Sample Viewing. Neoprene Pad and Rubber Mounting to Prevent Slippage
Catalog No. 50-153-2318 Supplier Chemglass Life Sciences Supplier No. CLS347801
Well Locator Stand. Works perfectly with printed well-orienters. Place your 96 well plate on this 30 angled platform for easier visibility. Convenient Stand to be Used with CLS-3477 Well Locators. Tilted at a 30 Angle for Easy Sample Viewing. Neoprene Pad and Rubber Mounting to Prevent Slippage