Learn More
Thomas Scientific 96 Well Half Skirt PCR Plate
SDP
Catalog No. NC9281177
Encompass_Preferred
For use w/ ABI 3730 and Most Cycling Systems. Certified RNase, DNase and Pyrogen Free.
Catalog No. NC9281177 Supplier Thomas Scientific Supplier No. 1148G62
For use w/ ABI 3730 and Most Cycling Systems. Certified RNase, DNase and Pyrogen Free.