Learn More
Andwin Scientific Well Plate Stand for Microplates
SDP
Catalog No. NC9693628
Encompass_Preferred
The Well Plate Stand is made to perform as a holder for microplates. This product is tilted at a 30 to permit users to conveniently look at samples and pipettes without strain. Each stand features a neoprene pad and rubber mounting to reduce the risk of slippage of the plate and stand.
Catalog No. NC9693628 Supplier Andwin Scientific Supplier No. WPST1000
The Well Plate Stand is made to perform as a holder for microplates. This product is tilted at a 30 to permit users to conveniently look at samples and pipettes without strain. Each stand features a neoprene pad and rubber mounting to reduce the risk of slippage of the plate and stand.