Learn More
Sigma Aldrich Fine Chemicals Biosciences Well plate stand adjustabl1EA
Catalog No. 502900588
Encompass
This device tips the Well Orienter card and the microplate 30 allowing the investigator to identify wells clearly whether sitting or standing. Comfortably view and pipet without strain. The Well Stand will free up one hand since you do not have to constantly tip the plate back and forth while pipetting.
Catalog No. 50-290-0588 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7435571EA
This device tips the Well Orienter card and the microplate 30 allowing the investigator to identify wells clearly whether sitting or standing. Comfortably view and pipet without strain. The Well Stand will free up one hand since you do not have to constantly tip the plate back and forth while pipetting.