Learn More
Sigma Aldrich Fine Chemicals Biosciences Well plate stand pkg of 11EA
Catalog No. 502900347
Encompass
This device tips the Well Orienter card and the microplate 30 allowing the investigator to identify wells clearly whether sitting or standing. Comfortably view and pipet without strain. The Well Stand will free up one hand since you do not have to constantly tip the plate back and forth while pipetting.
Catalog No. 50-290-0347 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W07651EA
This device tips the Well Orienter card and the microplate 30 allowing the investigator to identify wells clearly whether sitting or standing. Comfortably view and pipet without strain. The Well Stand will free up one hand since you do not have to constantly tip the plate back and forth while pipetting.