Learn More
World Precision Instrument Well-Style Reference Electrode for WPI MICRO-ePORE™
SDP
Catalog No. 502521213
Encompass
The reference electrode ring is secured to a microscope slide with a vacuum gel, and the media is placed inside the well-style electrode. Both the probe-style electrode and the well-style electrode are included with the MICRO-ePORE™ system. This is a replacement part. [Ring reference electrode – OD: 12.7 mm (0.5”), Thickness: 0.4 mm (0.16”)]
Catalog No. 50-252-1213 Supplier World Precision Instrument Supplier No. 99190
The reference electrode ring is secured to a microscope slide with a vacuum gel, and the media is placed inside the well-style electrode. Both the probe-style electrode and the well-style electrode are included with the MICRO-ePORE™ system. This is a replacement part. [Ring reference electrode – OD: 12.7 mm (0.5”), Thickness: 0.4 mm (0.16”)]