Learn More
Research Products International Corp Well-Align Pipetter Guide, for Flat Bottom Micro-Plates and Conical Bottom PCR Plates
SDP
Catalog No. 50550326
Encompass
Simply move the sliding guide across the plate to align the pipetter with the micro-plate wells. Feature permanent etched column letters for quick and easy orientation. Reduces eye fatigue and helps eliminate pipetting errors. Designed for flat bottom microplates and conical bottom PCR plates. Compatible with all single or multi-channel pipettes. Can be used with or without angled plate stand.
Catalog No. 50-550-326 Supplier Research Products International Corp Supplier No. 249905
Simply move the sliding guide across the plate to align the pipetter with the micro-plate wells. Feature permanent etched column letters for quick and easy orientation. Reduces eye fatigue and helps eliminate pipetting errors. Designed for flat bottom microplates and conical bottom PCR plates. Compatible with all single or multi-channel pipettes. Can be used with or without angled plate stand.