Learn More
Description
EDTA solution is intended for use with the Wellcogen™ Kits (ZL20/R30858701, ZL21/R30858801, ZL22/R30859001, ZL23/ R30859203, ZL24/R30859502 and ZL26/R30859602) as a diluent in which to heat serum samples for subsequent testing with the Wellcogen™ latex tests.
Intended for use as a diluent in which to heat serum samples for subsequent testing with Wellcogen latex tests R30858701, R30858801, R30859001, R30859203, R30859502 and R30859602.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Content And Storage | 2°C to 25°C |
| Description | Wellcogen EDTA Solution |
| CE Marker | Yes |
| Concentration | 0.1M |
| Format | Vial |
| Quantity | 10 mL/Vial |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.