Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Wellcogen™ Haemophilus influenzae b Rapid Latex Agglutination Test

Catalog No. R30858801 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
30 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R30858801 30 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R30858801 Supplier Thermo Scientific™ Supplier No. R30858801
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rapidly detect antigens to Haemophilus influenzae present in human body fluids or blood cultures by latex agglutination.

Wellcogen™ H. influenzae b is a rapid latex test for use in the qualitative detection of antigen from Haemophilus influenzae type b, present in body fluids as a consequence of infection or in blood cultures. NOTE: Tests performed directly on clinical specimens are intended for screening purposes and should augment, not replace, culture procedures. Results must be used in conjunction with other data; eg symptoms, results of other tests, clinical impressions etc

Use this rapid latex test in the qualitative detection of antigen from Haemophilus influenzae type b, present in body fluids as a consequence of infection, or in blood cultures.

The reagent consists of polystyrene latex particles which have been coated with antibodies specific to H. influenzae type b antigen. These latex particles agglutinate in the presence of sufficient homologous antigen.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Wellcogen Haemophilus influenzae b Test Kit
Detectable Analytes Influenza B
Sample Type Body Fluids, Blood Culture
Type Reagent
Quantity 30 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.