Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Wellcolex™ Color Salmonella Rapid Latex Agglutination Test Kit

Catalog No. R30858302
Change view
Click to view available options
Quantity:
200 Tests/Kit
50 Tests/Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R30858302 200 Tests/Kit
R30858301 50 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R30858302 Supplier Thermo Scientific™ Supplier No. R30858302
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Save up to two days over traditional screening methods with this easy-to-use, cost effective latex agglutination test for rapid, accurate identification of Salmonella.

Rapidly detect and presumptively identify Salmonella from culture by latex agglutination using Thermo Scientific™ Wellcolex™ Color Salmonella Rapid Latex Agglutination Test Kit. Save up to two days' testing time over traditional screening procedures with this easy-to-use, cost-effective test. With its high positive and negative predictive values (PPV > 98.7%) and results in less than three minutes, laboratories can use this rapid screening test to eliminate Salmonella-negative samples from further testing, reducing the number of samples requiring confirmatory testing.

Use this rapid latex agglutination test for detection and presumptive serogrouping of Salmonella from Selenite Broth and solid culture media. Wellcolex Color Salmonella can also be used for the presumptive identification of Salmonella serogroups A, B, C, D, E, and G, and the Vi antigen using just two reagents.

  • Identifies major groups of Salmonella
  • Utilizes advanced color technology for clear, easy-to-read species and/or serogroup identification
  • Provides accurate, reliable results in less than three minutes
  • Quickly eliminates negative samples when used as primary culture screening method

To perform Wellcolex Color Salmonella, a sample from a Selenite F broth culture (18 to 24 hours incubation) or a suspension of bacteria from solid agar is reacted with two test reagents, consisting of a mixture of suspensions of red, blue and green latex particles, each of which is coated with antibody specific for different Salmonella serogroups.

In the presence of homologous antigen one of the colors in the mixture will agglutinate, and the identity of the antigen is indicated by the color of the aggregated particles with a contrasting change in the color of the background. Each combination is easily distinguished from the others and from a negative result in which the particles remain in smooth grey‑brown suspension, and the occasional non‑specific result in which all the particles agglutinate into grey‑brown aggregates against a cleared background.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Wellcolex Colour Salmonella Kit
CE Marker Yes
Detectable Analytes Salmonella
Type Salmonella Test Kit
Includes Latex reagents 1 and 2, red, blue, and green positive controls, disposable mixing sticks, disposable reaction cards, disposable sample dispensers, instructions for use, and reading guide
Organism Group Group A, B, C, D, E and G
Quantity 200 Tests/Kit
Target Organism Class Salmonella
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.