Learn More
Research Products International Corp 96 Well Micro-Plate Guide, Horizontal Format, 2 per Package
SDP
Catalog No. 50550281
Encompass
Bright color lanes in horizontal orientation clearly define columns and rows on assay plates. Help reduce fatigue and errors when pipetting multiple samples into plates and improve efficiency. Reusable plastic cards fit most standard flat and round bottom plates.
Catalog No. 50-550-281 Supplier Research Products International Corp Supplier No. 247812
Bright color lanes in horizontal orientation clearly define columns and rows on assay plates. Help reduce fatigue and errors when pipetting multiple samples into plates and improve efficiency. Reusable plastic cards fit most standard flat and round bottom plates.