Learn More
MALIX LLC WELLlife COVID-19 / Flu A&B Quickly and accurately detect SARS-CoV-2, flu A, and flu B with a single nasal swab. Rapid results in 10 minutes.
Supplier:  MALIX LLC WELLLIFE
Encompass_Preferred
WELLlife COVID-19/Influenza A&B Test
The WELLlife™ COVID-19 / Influenza A&B Tests use advanced immunoassay technology to quickly and accurately differentiate between SARS-CoV-2, influenza A, and influenza B with a single nasal swab. Recommended for rapid diagnostics, these tests support swift treatment decisions during the flu season and amidst public health concerns. Pack of 25 Kits, 2 per Each Kit
Features and Benefits:
Cost-effective: Differentiates between SARS-CoV-2, influenza A, and influenza B antigens with a single test using an anterior nasal swab specimen.
Rapid Results: Available in 10 minutes, allowing for testing and treatment decision-making during the same office visit.
Quality Assurance: Outstanding clinical performance, including built-in kit controls for external quality testing, enables more confident decisions.
Catalog No. NC3235778