Learn More
Wells Lamont™ All Day Inspector Gloves
Catalog No. 19048693 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Glove Cuff Color:
Black
Gold
Purple
Size:
Small
Medium, Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Glove Cuff Color Size
19-048-693 Black X-Large
19-048-691 Purple Small
19-048-692 Gold Medium, Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-048-693 Supplier Wells Lamont™ Supplier No. 3111MDFPBXL
This item is not returnable. View return policy
Specifications
Description PHA Fiber
Laboratory/Cleanroom Use Yes
Material Nylon, PHA
Size X-Large
Thickness (English) 15 mil
Finger Style Full
For Use With (Application) To create a barrier for Latex, Rubber and Plastic gloves, used as a glove liner or alone
Glove Cuff Color Black