Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Whizard™ Cut Resistant Sleeve, Medium Duty

Catalog No. 1140024C Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Length (English):
22 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
11-400-24C 22 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-400-24C Supplier Wells Lamont™ Supplier No. 333482
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Whizard™ Medium Duty Cut Resistant Sleeves offer superior cut protection from arm lacerations. Constructed from a combination of high performance materials offering A7 ANSI cut protection. The medium duty armguard offers great protection for the arm and is available in different lengths.

  • Gray armguard provides strength, and good cut resistance while hiding dirt
  • Features High Performance Fibers and Stainless Steel
  • Outstanding wear for a long service life
  • ANSI Cut Level A7
  • Prewashed and preshrunk for a close fit
  • 20 in. and 22 in. has a clip attached on the upper arm that allows the sleeve to stay in place
  • Made in the USA

Specifications

Size Universal
Material Fiber, Stainless Steel
Cuff Style Knit
For Use With (Application) Glass handling, Metal fabrication, door and window manufacturing
Includes Clip
Length (English) 22 in.
Length (Metric) 55.8 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.