Learn More
MSC Wells Lamont Cut Abrasion Gloves, Cut A4, Small, 1 Pair
Catalog No. 5000792662
Encompass
These are cut and abrasion-resistant gloves. They are characterized by a Stainless Steel Fiber primary material, an uncoated coating, and come in a gray pair. The gloves are designed with an extended Kevlar thumb crotch, Aramid blend terry cloth, and a loop-out structure to provide cushioning for handling heavy parts. They also feature flame-resistant fibers for incidental spark protection.
  • Extended Kevlar thumb crotch
  • Aramid blend terry cloth
  • Loop-out structure provides a cushion to handle heavy parts
  • Flame-resistant fibers for incidental spark protection
Catalog No. 50-007-92662 Supplier MSC Supplier No. 80878440
These are cut and abrasion-resistant gloves. They are characterized by a Stainless Steel Fiber primary material, an uncoated coating, and come in a gray pair. The gloves are designed with an extended Kevlar thumb crotch, Aramid blend terry cloth, and a loop-out structure to provide cushioning for handling heavy parts. They also feature flame-resistant fibers for incidental spark protection.
  • Extended Kevlar thumb crotch
  • Aramid blend terry cloth
  • Loop-out structure provides a cushion to handle heavy parts
  • Flame-resistant fibers for incidental spark protection