Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Cut-Resistant FR Sleeves

Catalog No. 19148975 Shop All Wells Lamont Industry Group Products
Click to view available options
Length (English):
10 in.
14 in.
18 in.
24 in.
Style:
Sleeve with Thumb Hole
Standard Sleeve
This item is not returnable. View return policy
This item is not returnable. View return policy

Composite Cut-Resistant FR Sleeves offer a high level of cut protection, while also providing outstanding comfort and fit. Made of a combination of high performance materials and flame resistant fibers, which will allow it to be used where incidental protection against flame and sparks are required.

  • Unique knit sleeve made from a patented combination of high performance materials and flame resistant fibers
  • Knitted using strong para-aramid yarns
  • Lightweight construction will increase employee acceptance and reduce the risk of injury
  • Blousy fit allows superior comfort and breathability
  • Elastic on both ends allows sleeve to stay in place
  • Available in several lengths, with or without a thumb hole, and also a wide top design for larger biceps
  • ANSI Cut Level A3
  • ANSI Flame Rating 4
  • Sold by the dozen pieces
  • Made in the USA

Specifications

Product Type Cut-resistant Sleeve
Construction Lightweight high performance materials and flame resistant fiber
Color White, Yellow, Black
Style Sleeve with Thumb Hole
Closure Type Elastic Ends
Cut Resistance Rating ANSI Level 3
Cuff Style Knit
Cleaning Procedure Laundered using a mild soap and detergent, use a warm water wash with a temperature not to exceed 160°F, wash time should not be longer than 15 to 20 min, use a rinse cycle for the product in cold water, use a high extraction speed, tumb
For Use With (Application) Used to increase employee acceptance and reduce the risk of injury
Length (English) 24 in.
Length (Metric) 60.96 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.