Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Economy Grade Fleece Lined Grain Cowhide Leather Drivers Gloves

Catalog No. 19148828 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-148-828 Small
19-148-827 Medium
19-148-826 Large
19-148-825 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-148-828 Supplier Wells Lamont™ Supplier No. Y0062S
  • $321.50
    $220.65 / Pack of 12

    Save $100.85

    31% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cowhide Leather Drivers Gloves are lined with fleece to provide comfort and warmth in cold environments such as refrigeration and warehousing.

  • Grain cowhide leather construction with fleece lining
  • Shirred elastic back
  • Keystone thumb
  • Bound hem

Specifications

Color Natural
Grade Economy
Hand Specific
Material Grain Cowhide Leather
Product Type Driver Gloves
Size Small
Style Driver
For Use With (Application) Warehousing, Construction, Outdoors, Freezers
Gender Unisex
Glove Cuff Color Red
Glove Cuff Style Open
Hem Color Red
Liner Material Fleece
Palm Type Grain Cowhide
Series Y0062
Thumb Style Keystone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.