Learn More
MSC Wells Lamont FlexTech HPPE/PU Palm, ANSI A2, XLarge, 1 Pair
Catalog No. 5000793853
Encompass
The Y9265 FlexTech HPPE Shell with PU Palm glove provides a balance of comfort, dexterity, and cut resistance. It features high-performance polyethylene fibers (HPPE) and a gray nylon blend in its shell for enhanced cut protection. The polyurethane palm coating delivers excellent grip and tactility, making it suitable for general assembly and applications requiring robust cut protection.
  • FlexTech high performance polyethylene fibers (HPPE) and gray nylon blend shell
  • Polyurethane palm coating provides excellent grip and tactility
  • Launderable for an extended wear life
  • Features 13 gauge for optimal dexterity
  • ANSI/ISEA 105-2024 Cut Level A2
  • ANSI/ISEA 105-2024 Abrasion Level 6
  • ANSI/ISEA 105-2024 Puncture Level 4
  • Open, uncoated back design promotes air circulation
Catalog No. 50-007-93853 Supplier MSC Supplier No. 68870443
The Y9265 FlexTech HPPE Shell with PU Palm glove provides a balance of comfort, dexterity, and cut resistance. It features high-performance polyethylene fibers (HPPE) and a gray nylon blend in its shell for enhanced cut protection. The polyurethane palm coating delivers excellent grip and tactility, making it suitable for general assembly and applications requiring robust cut protection.
  • FlexTech high performance polyethylene fibers (HPPE) and gray nylon blend shell
  • Polyurethane palm coating provides excellent grip and tactility
  • Launderable for an extended wear life
  • Features 13 gauge for optimal dexterity
  • ANSI/ISEA 105-2024 Cut Level A2
  • ANSI/ISEA 105-2024 Abrasion Level 6
  • ANSI/ISEA 105-2024 Puncture Level 4
  • Open, uncoated back design promotes air circulation