Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ FlexTech™ Full NBR Coated Synthetic Knit Gloves with Nitrile Coated Palm

Catalog No. 17200327 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-200-327 X-Small
17-200-328 Small
17-200-329 Medium
17-200-330 Large
17-200-331 X-Large
17-200-332 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-200-327 Supplier Wells Lamont™ Supplier No. Y9289XS
Add to Cart
Add to Cart
This item is not returnable. View return policy

FlexTech™ Full NBR Coated Synthetic Knit Gloves with Nitrile Coated Palm are lightweight, synthetic knit shells that are double coated with a layer of nitrile rubber (NBR) for water-resistance, and an additional layer of nitrile coating on the palm and fingers for enhanced grip capabilities.

  • Lightweight, synthetic knit shell with double coating
  • Sandy nitrile coated palm and fingertips enhances grip
  • Full NBR coating provides an additional layer of protection against liquids
  • Gloves offer a balance of comfort and dexterity

Specifications

ANSI Abrasion Rating ANSI 4
Coated Area Palm, Fingers
Coating Material Nitrile
Color Blue
Cut Resistance Rating ANSI A1
Exterior Finish Fully Textured
Hand Specific
Latex Free Yes
Material Nitrile Rubber
Product Type Dip-coated Gloves
Size X-Small
For Use With (Application) General Assembly, Small Parts Handling, Painting, and Manufacturing
Glove Cuff Color Gray
Glove Cuff Style Knit
Series Y9289
Yarn Gauge 13
Length (English) 10 in.
Length (Metric) 25.4 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.