Learn More
MSC Wells Lamont Glove, ANSI A5, Left Hand, Large, 1 ea
Catalog No. 5000792730
Encompass
These cut resistant gloves are designed to provide hand protection in demanding industrial environments. Featuring a Spectra/stainless steel liner and urethane pattern, they offer excellent resistance to cuts and abrasions.
  • Cut resistant design for enhanced safety
  • Spectra/stainless steel liner for superior durability
  • Urethane pattern for improved grip and handling
  • Knitwrist cuff ensures a comfortable and secure fit
  • Meets ANSI Cut Level A5 for high-level protection
  • Suitable for steel processing, metal handling, and blade handling applications
  • Detailed laundering instructions for extended glove life
Catalog No. 50-007-92730 Supplier MSC Supplier No. 86518891
These cut resistant gloves are designed to provide hand protection in demanding industrial environments. Featuring a Spectra/stainless steel liner and urethane pattern, they offer excellent resistance to cuts and abrasions.
  • Cut resistant design for enhanced safety
  • Spectra/stainless steel liner for superior durability
  • Urethane pattern for improved grip and handling
  • Knitwrist cuff ensures a comfortable and secure fit
  • Meets ANSI Cut Level A5 for high-level protection
  • Suitable for steel processing, metal handling, and blade handling applications
  • Detailed laundering instructions for extended glove life