Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wells Lamont™ GuardTec3™ Hi-Vis Foam Latex Palm Gloves
Shop All Wells Lamont Industry Group ProductsSpecifications
Specifications
| Color | Gray, Orange |
| Cut Resistance Rating | ANSI A3 |
| Material | Nitrile, FlexTech |
| Product Type | Cut-resistant Gloves |
| Size | 2X-Large |
| For Use With (Application) | Cut protection, high visibility applications |
| Glove Cuff Color | Black |
| Glove Cuff Style | Knit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?