Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Extra Heavyweight Cut and Sewn Terrycloth High Temperature Gloves

Catalog No. 19013272 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-013272 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-013272 Supplier Wells Lamont™ Supplier No. Y946
  • $124.52
    $94.65 / Pack of 6

    Save $29.87

    24% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extra Heavyweight Cut and Sewn Terrycloth High Temperature Gloves with loop-out terrycloth creates an insulating barrier between a hot object and the user for protection from temperatures up to 350°F.

  • Exta heavyweight cut and sewn construction
  • Loop-out terrycloth increases heat resistance
  • Gauntlet cuff extends protection up the arm to prevent burns
  • Provides protection from temperatures up to 350°F
  • Ambidextrous

Specifications

Color White
Hand Ambidextrous
Material Terrycloth
Product Type High Temperature Gloves
Size X-Large
Temperature (English) Max. 350°F
Temperature (Metric) Max 177°C
For Use With (Application) Steel Processing
Glove Cuff Style Gauntlet
Series Y946
Length (English) Cuff 5 in.
Length (Metric) Cuff 12.7 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.