Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Lightweight Cotton Lisle Inspection Gloves

Catalog No. 1146226A Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Gender:
Men's
Women's
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Gender
11-462-26A Women's
11-462-26B Men's
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 11-462-26A Supplier Wells Lamont™ Supplier No. Y6701W
  • $39.25
    $34.65 / Dozen of 12

    Save $4.60

    12% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lightweight Cotton Lisle Inspection Gloves feature a form-fitting, two-piece design constructed of 100% cotton to provide comfort and absorb perspiration when used as a glove liner underneath liquid-proof gloves.

  • 100% cotton lisle
  • Form-fitting, two-piece construction
  • Can be worn alone or used as a glove liner
  • Unhemmed cuff
  • Moisture absorbing

Specifications

Construction Cut and Sewn
Coating Material Uncoated
Color White
Disposable Single-use
Hand Ambidextrous
Laboratory/Cleanroom Use Yes
Material Cotton
Product Type Inspection Gloves
For Use With (Application) Electronics Assembly, Industrial Manufacturing
Gender Women's
Glove Cuff Style Plain
Series Y6701
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.