Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Jomac™ Standard Weight Cotton/Polyester Gloves

Catalog No. 114626 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
11-462-6 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-462-6 Supplier Wells Lamont™ Supplier No. 1666
  • $132.00
    $93.65 / Dozen of 12

    Save $38.35

    29% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Jomac™ Standard Weight Cotton/Polyester Gloves with loop out terrycloth construction provide protection from temperatures up to 300°F.

  • White loop-out terrycloth construction
  • Protection from temperatures up to 300°F
  • Knit wrist helps prevent dirt and debris from entering the glove

Specifications

Color White
Cut Resistance Rating ANSI A2
Hand Ambidextrous
Material Terrycloth
Product Type Cotton/Polyester Gloves
Size Large
Temperature (English) Max. 300°F
Temperature (Metric) Max 145°C
For Use With (Application) Metal Handling, Steel Processing
Glove Cuff Style Knit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.