Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Jomac™ Heavyweight Terrycloth Heat Resistant Sleeves

Catalog No. 19013507 Shop All Wells Lamont Industry Group Products
This item is not returnable. View return policy
Catalog No. 19-013507 Supplier Wells Lamont™ Supplier No. S15HR
Add to Cart
Add to Cart
This item is not returnable. View return policy

Jomac™ Heavyweight Terrycloth Heat Resistant Sleeves with loop out terrycloth construction provide protection from temperatures up to 350°F.

  • Protection from temperatures up to 350°C
  • Elastic top secures sleeve to upper arm
  • Knit wrist helps prevent dirt and debris from entering the sleeve

Specifications

Material Terrycloth
Color White
Disposable Reusable
Closure Type Elastic Upper with Knit Cuff
Cuff Style Knit
For Use With (Application) Ideal for baking or working around ovens.
Length (Metric) 35.56 cm
Temperature (English) Max. 662°F
Temperature (Metric) Max 350°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.