Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ MechPro™ 7750 Synthetic Leather Gloves with Thinsulate™ Lining

Catalog No. 19148761 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-148-761 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-148-761 Supplier Wells Lamont™ Supplier No. 7750XL
  • $514.00
    $234.65 / Dozen of 12

    Save $279.35

    54% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $2,678.00
    $1,224.65 / Case of 6 DZ

    Save $1,453.35

    54% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MechPro™ 7750 Synthetic Leather Gloves with Thinsulate™ Lining offer protection from nicks, abrasion, and cold weather conditions. Ideal for enhancing grip and dexterity when handling tools or small parts.

  • Synthetic leather palm and fingertips
  • Ribbed knit fabric on back of hand
  • Reinforced fingertips with improved pinched fingertip design
  • G80 Thinsulate™ lining for cold weather protection
  • Feeltite™ hook and loop wrist closure
  • Machine washable

Specifications

Color Black
Hand Specific
Material Synthetic Leather
Product Type Insulated Gloves
Size X-Large
For Use With (Application) Construction, Mechanics, Steel/Metal Handling, and Warehousing
Glove Cuff Style Hook and Loop
Liner Material Thinsulate™
Lining Thinsulate™
Series 7750
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.