Learn More
Wells Lamont™ Grain Cowhide Leather MIG/TIG Welding Gloves
Catalog No. 19148752 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Large
Catalog No. Size
19-148-752 Large
1 options
This item is not returnable. View return policy
Catalog No. 19-148-752 Supplier Wells Lamont™ Supplier No. Y2021L
Add to Cart
Add to Cart
This item is not returnable. View return policy
Grain Cowhide Leather MIG/TIG Welding Gloves feature grain cowhide leather construction that is durable, with Aramid Fiber threads for increased strength and burn resistance.
  • Quality grain cowhide leather construction with a split leather cuff
  • Cowhide leather has natural heat and abrasion resistance
  • Sewn with Aramid Fiber for increased strength and burn resistance
  • Ideal for welding applications in various industries such as automotive, construction, and sheet metal
Specifications
Cuff Material Split Leather
Coating Material Uncoated
Cut Resistance Rating ANSI A1
Material Grain Cowhide Leather, Aramid Fiber
Product Type MIG/TIG Welding Gloves
Size Large
Style Gunn
For Use With (Application) MIG/TIG Welding
Glove Cuff Color Gold
Glove Cuff Style Gauntlet
Hem Color Brown
Puncture Rating ANSI/ISEA 105-16 Level 4
Series Y2021
Thumb Style Wing
Length (English) Cuff 4 in.
Length (Metric) Cuff 10 cm