Learn More
Wells Lamont™ PERM-RUFF Red PVC Gloves
Catalog No. 19076332 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Large
Catalog No.
19-076332 Large
1 options
This item is not returnable. View return policy
Catalog No. 19-076332 Supplier Wells Lamont™ Supplier No. PR4114L
This item is not returnable. View return policy
Specifications
Color Red
Material PVC
Size Large
For Use With (Application) Lumber handling, Petrochemical, refining, handling abrasive materials coated with acids, caustics, solvents, grease and oily environments
Glove Cuff Style Gauntlet
Liner Material Interlock Knit
Lining Lined
Length (English) 14 in.
Length (Metric) 35.5 cm