Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Select Grade Shoulder Split Cowhide Leather Palm Work Gloves

Catalog No. 19823913 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Medium
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-823-913 Medium
19-823-912 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-823-913 Supplier Wells Lamont™ Supplier No. Y3107M
  • $140.36
    $84.65 / Dozen of 12

    Save $55.71

    40% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Select Grade Shoulder Split Cowhide Leather Palm Work Gloves are durable gloves featuring split leather index fingers, leather tips, and a leather knuckle strap. The lined palm provides additional comfort in the palm of the hand.

  • Select grade shoulder split cowhide leather
  • Striped canvas back and cuff
  • Full leather palm, fingertips, and knuckle strap
  • Gauntlet cuff
  • Wing thumb
  • Lined palm

Specifications

Construction Cut and Sewn
Cuff Material Canvas
Color Gray, Blue
Grade Select
Hand Specific
Material Shoulder Split Cowhide Leather
Product Type General Purpose Gloves
Size Medium
Style Gunn
For Use With (Application) Steel Processing, Metal Handling, Construction
Gender Unisex
Glove Cuff Color Blue with Yellow Stripes
Glove Cuff Style Gauntlet
Grip Design Smooth
Hem Color Green
Liner Material Fleece
Lining Lined
Palm Type Lined
Series Y3107
Thumb Style Wing
Length (English) Cuff 4 in.
Length (Metric) Cuff 10.2 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.