Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Reusable Half-Finger Knit Nylon Glove Liners

Catalog No. 189994591A Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Universal
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-999-4591A Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-999-4591A Supplier Wells Lamont™ Supplier No. M088
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reusable Half-Finger Knit Nylon Glove Liners are constructed of high-quality, 100% continuous filament nylon with a half finger design for increased tactile sensitivity. Wear alone, or under nitrile gloves to help reduce perspiration.

  • Constructed of high-quality, 100% continuous filament nylon
  • Half-finger design increases tactile sensitivity
  • Form fitting for high-dexterity applications
  • Wear alone, or under nitrile gloves to help prevent perspiration
  • Universal size easily accommodates various hand sizes
  • Made in US

Specifications

Coating Material Uncoated
Color White
Disposable Reusable
Hand Ambidextrous
Laboratory/Cleanroom Use Yes
Material Nylon
Product Type Glove Liners
Size Universal
Finger Style Half
For Use With (Application) Critical Environments, Cleanrooms, Electronics Assembly, and Inspection
Glove Cuff Style Knit
Series M08
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.