Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Reusable Half-Finger Nylon Glove Liners

Catalog No. 19086525 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Gender:
Men's
Women's
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Gender
19-086-525 Men's
19-086-524 Women's
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-086-525 Supplier Wells Lamont™ Supplier No. M006M.WLC
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reusable Half-Finger Nylon Glove Liners are constructed of 100% nylon for a high level of mobility and comfort. Capable of withstanding up to 30 wash cycles without shrinkage.

  • 100% continuous nylon filament construction is thin and comfortable
  • Fully launderable up to 30 times without shrinkage
  • Reduced replacement costs and waste disposal
  • Ideal for use in critical environments and clean rooms
  • Half-finger length
  • Ambidextrous
  • Made in US

Specifications

Coating Material Uncoated
Color White
Disposable Reusable
Hand Ambidextrous
Laboratory/Cleanroom Use Yes
Material Nylon
Product Type Glove Liners
Finger Style Half
For Use With (Application) Critical Environments, Cleanrooms, Electronics Assembly, and Inspection
Gender Men's
Glove Cuff Style Knit
Hem Color Black
Series M006.LC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.