Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Select Shoulder Split Leather Palm Work Gloves

Catalog No. 19148812 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Color:
Gray, Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-148-812 Gray, Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-148-812 Supplier Wells Lamont™ Supplier No. Y3106S
This item is not returnable. View return policy

Gloves feature split leather on the palm and index fingers with a leather knuckle strap on the back of the hand. Lined palm provides additional comfort in the palm of the hand.

  • Durable, shoulder split leather construction
  • Striped canvas back and cuff for breathability and comfort
  • Full leather palm, fingers, and knuckle strap
  • Safety cuff
  • Wing thumb

Specifications

Color Gray, Blue
Grade Select
Hand Specific
Material Shoulder Split Cowhide Leather
Product Type General Purpose Gloves
Size Small
Glove Cuff Style Safety
Hem Color Red
Liner Material Cotton
Thumb Style Wing
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.