Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Spectra Cut-Resistant Glove Liners

Catalog No. 19050034A Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Hand:
Ambidextrous
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Hand
19-050-034A Ambidextrous
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-050-034A Supplier Wells Lamont™ Supplier No. 135259
This item is not returnable. View return policy

Specifications

Certifications/Compliance Meet ANSI Cut Level A1 (ASTM F2992-15) requirements.
Color White
Cut Resistance Rating ANSI A4
Hand Ambidextrous
Material Fibers
Product Type Cut-resistant Gloves
Size Small
For Use With (Application) Cut and abrasion resistance
Glove Cuff Color Yellow
Glove Cuff Style Knit
Hem Color Yellow
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.