Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Seamless Knit Thermal Liner - 13 Gauge

Catalog No. 19821185 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Universal
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-821-185 Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-821-185 Supplier Wells Lamont™ Supplier No. Y1451
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Seamless Knit Thermal Liner is constructed of a polyester/Lycra™ blend to wick away moisture, and keep hands warm. Gloves can be worn alone, or used as a liner in cold weather applications.

  • Comfort combined with thermal protection
  • Glove composition allows for quick drying to keep hands warm
  • Can be worn alone, or used as a glove liner
  • Made in the USA

Specifications

Coating Material Uncoated
Color White
Disposable Reusable
Hand Ambidextrous
Material Polyester, Lycra
Product Type Thermal Liners
Size Universal
For Use With (Application) Thermal Protection
Glove Cuff Color White
Glove Cuff Style Knit
Series Y1451
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.