Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Whizard™ Cut-Tec™ Spectra™ Composite Fiber Gloves

Catalog No. 17985543 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Cut Resistance Rating:
ANSI A2
Size:
Small
Medium
Glove Cuff Color:
Blue
Yellow
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cut Resistance Rating Size Glove Cuff Color
17-985-543 ANSI A2 Medium Blue
17-985-542 ANSI A2 Small Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-985-543 Supplier Wells Lamont™ Supplier No. Y5858M
This item is not returnable. View return policy

Specifications

Color White
Cut Resistance Rating ANSI A2
Hand Ambidextrous
Material Composite Fiber, Lycra
Product Type Cut-resistant Gloves
Size Medium
For Use With (Application) Provides durability and cut protection
Glove Cuff Color Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.