Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Whizard™ Cut-Resistant Glove Liners
GSA_VA

Catalog No. 19050036C Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-050-036C Large
19-050-036A Small
19-050-036B Medium
19-050-036D X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-050-036C Supplier Wells Lamont™ Supplier No. M281L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Whizard™ Cut-Resistant Glove Liners featuring a stainless steel and polyester construction provide resistance to cuts and abrasions, while dark coloring helps to conceal dirt. Wear alone, or under another pair of gloves for additional protection.

  • Glove liners conceal dirt and provide ANSI Cut Level A4 protection
  • Wear alone, or under another pair of gloves for additional protection
  • Moderate tactile sensitivity
  • Latex-free
  • Category III for particulates
  • Made in the USA

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Coating Material Uncoated
Color Black
Cut Resistance Rating ANSI A4
Disposable Reusable
Hand Ambidextrous
Latex Free Yes
Material Stainless Steel, Polyester
Product Type Cut-resistant Glove Liners
Size Large
For Use With (Application) Critical Environments, Cleanrooms, and Electronics Assembly
Glove Cuff Style Knit
Hem Color Blue
Series M281
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.