Learn More
THERMO SCI LSG 3P Thermo Scientific Wellwash Microplate Washer, 100/240 V, 1 ea
Supplier:  THERMO SCI LSG 3P 5165000
Encompass_Preferred
The Wellwash Microplate Washer is an easy-to-use microplate-strip washer designed for routine ELISA applications, capable of washing 96-well plates securely. It offers reliable performance and versatility, featuring a graphical interface and a USB port for convenience.
  • Easy and convenient to use with large color display and keypad.
  • Logical and intuitive user interface simplifies protocol development.
  • Reliable and secure performance with unpressurized bottles and liquid level sensors.
  • Automatic rinse and prime features, and an aerosol cover.
  • Optimal washing performance with sweep mode for low residual volume.
  • Adjustable dispense and aspiration heights, and aspiration speed.
  • USB port for protocol transfer and report printing.
  • Includes a 1x8 wash head.
Catalog No. 50-190-3757
Add to Cart
Request a Quote
Description
Description
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov