Learn More
THERMO SCI LSG 3P Thermo Scientific Wellwash Microplate Washer, 1x12, 96-Well, 1 ea
Supplier: THERMO SCI LSG 3P 5165040
The Wellwash Microplate Washer is a reliable instrument for washing 96-well plates in routine ELISA-based assays, featuring an intuitive graphical interface and robust design for consistent, reliable results.
- Large color LCD and keypad for easy operation
- Logic-driven interface simplifies protocol development
- Unpressurized bottles with liquid level sensors for safe use
- Automatic prime feature for consistent performance
- Aerosol cover minimizes aerosolization risk
- Precise washing with <5% accuracy, <3% CV at 300 µL
- Low residual volume (<2 µL) for reproducible assay results
- Lightweight (8 kg) and compact design for lab spaces
- USB port for protocol transfer and lab integration
- Includes 1x12 wash head, 2 L wash bottle, 2 L waste bottle
Description
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.