Learn More
MSC Werner Climbing Harness, XL, Back and Front D-Rings, 400 lb
Catalog No. 5000765567
Encompass
The Werner Blue Armor H123004 Climbing Harness is designed with Relief Handles to mitigate suspension trauma, featuring WebAlert™ inspectable webbing, impact indicators, and 5-Point adjustments for a secure, visible, and comfortable fit.
  • Built-in relief handles provide immediate relief after a fall arrest
  • WebAlert™ inspectable webbing highlights wear and damage for easy inspection
  • Lightweight, form-fitting padding with a breathable compression interior protects the user and prevents tangling
  • Front D-ring for climbing applications
  • Quick Connect buckles on chest and legs for fast, easy connections
  • Optimal fit is achieved with 5-point adjustments
  • Meets ANSI Z359.11 Standards for full body harnesses
Catalog No. 50-007-65567 Supplier MSC Supplier No. 37537602
The Werner Blue Armor H123004 Climbing Harness is designed with Relief Handles to mitigate suspension trauma, featuring WebAlert™ inspectable webbing, impact indicators, and 5-Point adjustments for a secure, visible, and comfortable fit.
  • Built-in relief handles provide immediate relief after a fall arrest
  • WebAlert™ inspectable webbing highlights wear and damage for easy inspection
  • Lightweight, form-fitting padding with a breathable compression interior protects the user and prevents tangling
  • Front D-ring for climbing applications
  • Quick Connect buckles on chest and legs for fast, easy connections
  • Optimal fit is achieved with 5-point adjustments
  • Meets ANSI Z359.11 Standards for full body harnesses