Learn More
MSC Werner Harness, Standard Back D-Ring, M/L, Polyester, 1 ea
Catalog No. 5000677181
Encompass
This fall protection harness offers immediate relief post-fall and features WebAlert™ inspectable webbing for safety. It provides an optimal fit and durability for industrial applications.
  • Built-in relief handles provide immediate post-fall relief, allowing a gravity override seated position.
  • Inspectable webbing highlights wear and damage, with multiple impact indicators for easier inspection.
  • Lightweight, form-fitting padding prevents tangling during donning and protects the user.
  • Slotted pass-through buckles on the chest strap fasten without twisting.
  • Tongue buckle leg straps are ideal for multiple users.
  • Ensures optimal fit with 5-point adjustments.
  • Meets ANSI Z359.11 Standards for full body harnesses.
Catalog No. 50-006-77181 Supplier MSC Supplier No. 20700985
This fall protection harness offers immediate relief post-fall and features WebAlert™ inspectable webbing for safety. It provides an optimal fit and durability for industrial applications.
  • Built-in relief handles provide immediate post-fall relief, allowing a gravity override seated position.
  • Inspectable webbing highlights wear and damage, with multiple impact indicators for easier inspection.
  • Lightweight, form-fitting padding prevents tangling during donning and protects the user.
  • Slotted pass-through buckles on the chest strap fasten without twisting.
  • Tongue buckle leg straps are ideal for multiple users.
  • Ensures optimal fit with 5-point adjustments.
  • Meets ANSI Z359.11 Standards for full body harnesses.