Learn More
MSC Werner Harness, Standard Back D-Ring, M/L Size, 1 ea
Catalog No. 5000795265
Encompass
This harness features Relief Handles to aid in achieving the Gravity Override position, reducing suspension trauma. It includes inspectable webbing and impact indicators for easier inspection. With 5-point adjustments, lightweight hardware, and bright colors, it enhances worker visibility and ensures an optimal fit. The rear dorsal D-ring is made from high-quality forged alloy steel for strength. A Quick Connect chest buckle and Tongue buckle leg straps allow for simple donning and doffing.
  • Built-in relief handles for post-fall relief
  • WebAlert™ inspectable webbing and impact indicators for easy inspection
  • Lightweight, breathable padding prevents tangling
  • Quick connect chest buckle
  • Tongue buckle leg straps suitable for multiple users
  • Five-point adjustments for optimal fit
  • Meets ANSI Z359.11 Standards
Catalog No. 50-007-95265 Supplier MSC Supplier No. 28475044
This harness features Relief Handles to aid in achieving the Gravity Override position, reducing suspension trauma. It includes inspectable webbing and impact indicators for easier inspection. With 5-point adjustments, lightweight hardware, and bright colors, it enhances worker visibility and ensures an optimal fit. The rear dorsal D-ring is made from high-quality forged alloy steel for strength. A Quick Connect chest buckle and Tongue buckle leg straps allow for simple donning and doffing.
  • Built-in relief handles for post-fall relief
  • WebAlert™ inspectable webbing and impact indicators for easy inspection
  • Lightweight, breathable padding prevents tangling
  • Quick connect chest buckle
  • Tongue buckle leg straps suitable for multiple users
  • Five-point adjustments for optimal fit
  • Meets ANSI Z359.11 Standards