Learn More
Cayman Chemical Werner Syndrome RecQ Helicase-IN-2, Purity: ≥98%, CAS: 2869954-51-6, 1 mg
SDP
Catalog No. 502943346
Encompass_Preferred
An inhibitor of WRN (IC50 = 100 nM); selectively inhibits the growth of SW48 cells over DLD-1-WRN-KO cells (GI50s = 100 and >10,000 nM, respectively); reduces tumor volume in an SW48 mouse xenograft model at 120 mg/kg per day
Catalog No. 50-294-3346 Supplier Cayman Chemical Supplier No. 416781 MG
An inhibitor of WRN (IC50 = 100 nM); selectively inhibits the growth of SW48 cells over DLD-1-WRN-KO cells (GI50s = 100 and >10,000 nM, respectively); reduces tumor volume in an SW48 mouse xenograft model at 120 mg/kg per day