Learn More
Cayman Chemical Werner Syndrome RecQ Helicase-IN-4, Purity: ≥95%, CAS: 2869954-53-8, 5 mg
SDP
Catalog No. 502943838
Encompass_Preferred
An inhibitor of WRN (IC50 = 60 nM); selectively inhibits the growth of SW48 cells over DLD-1-WRN-KO cells (GI50s = 70 and >10,000 nM, respectively); reduces tumor volume in an SW48 mouse xenograft model at 240 mg/kg per day
Catalog No. 50-294-3838 Supplier Cayman Chemical Supplier No. 416795 MG
An inhibitor of WRN (IC50 = 60 nM); selectively inhibits the growth of SW48 cells over DLD-1-WRN-KO cells (GI50s = 70 and >10,000 nM, respectively); reduces tumor volume in an SW48 mouse xenograft model at 240 mg/kg per day