Learn More
Wesco Industrial Products 50BT Series Drum Truck
Catalog No. NC1271652 Shop All Wesco Industrial Products Products
Encompass_Preferred
Poly Drum Truck & DIspensing 50BT Moldon Rubber Wheels Acts as a Poly Drum Truck or Drain Stand. Wide “feet” and belt eliminate the risk of drums being punctured by nose prongs and chime hooks. With two wheels resting on floor, 32" to center of belt. Handles all poly drums except overpacks). Handles 30, 55 & 85 gal. drums. Steel, poly & fiber drums. Capacity 1000 lbs. Weight 65 lbs.
Catalog No. NC1271652 Supplier Wesco Industrial Products Supplier No. 240083
Poly Drum Truck & DIspensing 50BT Moldon Rubber Wheels Acts as a Poly Drum Truck or Drain Stand. Wide “feet” and belt eliminate the risk of drums being punctured by nose prongs and chime hooks. With two wheels resting on floor, 32" to center of belt. Handles all poly drums except overpacks). Handles 30, 55 & 85 gal. drums. Steel, poly & fiber drums. Capacity 1000 lbs. Weight 65 lbs.