Learn More
MSC Wesco Industrial Products Drum Deheader Blade, Beryllium Alloy, 1 ea
Catalog No. 5000796113
Encompass
This drum deheader replacement blade is designed for use with non-sparking drum deheaders. It is constructed from beryllium alloy, ensuring safety in environments where sparks could be hazardous.
  • Constructed from beryllium alloy
  • Designed for non-sparking drum deheaders
  • Provides a reliable replacement for damaged blades
Catalog No. 50-007-96113 Supplier MSC Supplier No. 03093259
This drum deheader replacement blade is designed for use with non-sparking drum deheaders. It is constructed from beryllium alloy, ensuring safety in environments where sparks could be hazardous.
  • Constructed from beryllium alloy
  • Designed for non-sparking drum deheaders
  • Provides a reliable replacement for damaged blades