Learn More
Chemglass Life Sciences Condenser, West, 430mm, 24/40 Joint, 250mm Jacket Length
SDP
Catalog No. CG122003
Encompass_Preferred
Condenser, West, 430mm, 24/40 Joint, 250mm Jacket Length. Narrow annular space between jacket and inner tube of the West condenser provides for excellent cooling efficiency due to the increased flow rate of water. Condenser has a top standard taper outer joint and a lower inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration
Catalog No. CG122003 Supplier Chemglass Life Sciences Supplier No. CG122003
Condenser, West, 430mm, 24/40 Joint, 250mm Jacket Length. Narrow annular space between jacket and inner tube of the West condenser provides for excellent cooling efficiency due to the increased flow rate of water. Condenser has a top standard taper outer joint and a lower inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration