Learn More
Bioworld WEST NILE ENVELOPE VIRUS (R
Supplier:  Bioworld 220607561
Encompass_Preferred
WEST NILE ENVELOPE VIRUS (RECOMBINANT): Recombinant West Nile Envelope Virus Source: E.Coli Description : E.coli derived recombinant. The 18kda protein contains the West-Nile N-Terminal Envelope Virus immunodominant regions. The prot...
Catalog No. 50-253-857